missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Octadecanophenone, 99%

Manufacturer:  Alfa Aesar™ H5339414

 View more versions of this product

Catalog No. AAH5339414



Specifications

Specifications

n-Octadecanophenone
65°C to 67°C
6786-36-3
MFCD00010700
DBLXXVQTWJFJFI-UHFFFAOYSA-N
1-phenyloctadecan-1-one
81244
99%
425°C
25g
C24H40O
octadecanophenone, stearophenone, n-octadecanophenone, heptadecyl phenyl ketone, octadecanophenone, octadecanophenone, 1-octadecanone, 1-phenyl, 1-phenyl-1-octadecanone #
CCCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1
344.583
344.58

Chemical Identifiers

6786-36-3
344.583
DBLXXVQTWJFJFI-UHFFFAOYSA-N
81244
CCCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1
C24H40O
MFCD00010700
octadecanophenone, stearophenone, n-octadecanophenone, heptadecyl phenyl ketone, octadecanophenone, octadecanophenone, 1-octadecanone, 1-phenyl, 1-phenyl-1-octadecanone #
1-phenyloctadecan-1-one
SDS Documents