missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Octadecane, tech., 90%

Manufacturer:  Alfa Aesar™ 44301A1

 View more versions of this product

Catalog No. AA44301A1



Specifications

Specifications

n-Octadecane
166°C (330°F)
1.439
White
1kg
C18H38
ccris 681
RZJRJXONCZWCBN-UHFFFAOYSA-N
octadecane
11635
254.5
90%
0.782 g/mL
Odorless
317°C
28°C
593-45-3
MFCD00009007
Miscible with alcohol,acetone,ether and petroleum ether. Immiscible with water.
CCCCCCCCCCCCCCCCCC
254.502
CHEBI:32926
Crystalline Mass/Melt
Technical

Chemical Identifiers

593-45-3
254.502
RZJRJXONCZWCBN-UHFFFAOYSA-N
11635
octadecane
C18H38
MFCD00009007
ccris 681
CHEBI:32926
CCCCCCCCCCCCCCCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H304-H315-H319-H335

EINECSNumber : 209-790-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents