missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Octadecane, 99.5+%

Manufacturer:  Alfa Aesar™ 4451203

 View more versions of this product

Catalog No. AA4451203



Specifications

Specifications

n-Octadecane
166°C (330°F)
1.4390
White
1g
C18H38
ccris 681
CCCCCCCCCCCCCCCCCC
254.502
CHEBI:32926
Crystalline Mass/Melt
0.782g/mL
Odorless
317°C
28°C
593-45-3
MFCD00009007
RZJRJXONCZWCBN-UHFFFAOYSA-N
octadecane
11635
254.5
≥99.5%

Chemical Identifiers

593-45-3
254.502
RZJRJXONCZWCBN-UHFFFAOYSA-N
11635
octadecane
C18H38
MFCD00009007
ccris 681
CHEBI:32926
CCCCCCCCCCCCCCCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents