Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Octadecane, 99.5+%

$30.45 - $30.45

Chemical Identifiers

Chemical Name or Material n-Octadecane
CAS 593-45-3
Molecular Formula CH3(CH2)16CH3
Formula Weight 254.5
MDL Number MFCD00009007
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4451203

  SDS
alfa aesar™
4451203
1g Each for $30.45
Description & Specifications

Specifications

Physical Form Crystalline Mass/Melt
Formula Weight 254.5
Chemical Name or Material n-Octadecane
Refractive Index 1.4390
TSCA TSCA
Density 0.782g/mL
Melting Point 28°C
Boiling Point 317°C
Color White
Assay ≥99.5%
Quantity 1g
Flash Point 166°C (330°F)
Molecular Formula CH3(CH2)16CH3
Odor Odorless
MDL Number MFCD00009007
CAS 593-45-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.