Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(o-Tolyl)thiourea, 97+%

Chemical Identifiers

Chemical Name or Material N-(o-Tolyl)thiourea
CAS 614-78-8
Molecular Formula C8H10N2S
Formula Weight 166.25
MDL Number MFCD00041162
UN Number UN2811
Beilstein 2086802
Synonym N-(2-Methylphenyl)thiourea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2235009

  SDS
alfa aesar™
B2235009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2235018

  SDS
alfa aesar™
B2235018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2235030

  SDS
alfa aesar™
B2235030
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.25
Melting Point 155° to 158°C
TSCA TSCA
Synonym N-(2-Methylphenyl)thiourea
Quantity 10g
Chemical Name or Material N-(o-Tolyl)thiourea
CAS 614-78-8
Beilstein 2086802
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Odor Odorless
MDL Number MFCD00041162
UN Number UN2811
Health Hazard 1 Danger
Molecular Formula C8H10N2S
RTECS Number YU2975000
Assay ≥97%
Health Hazard 3 GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.