missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,O-Dimethylhydroxylamine hydrochloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 116340050
Description
Coupling reagentSpecifications
(10% aq. soln.) clear | |
Authentic | |
0.5% max. | |
112.0°C to 116.0°C | |
98% | |
97.5 | |
98% | |
CH3ONHCH3·HCl | |
04, 534 | |
methanamine, n-methoxy-, hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylamine hcl, n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride, n-methoxymethanamine hydrochloride, n-methoxymethylamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, o,n-dimethylhydroxylamine hydrochloride | |
USZLCYNVCCDPLQ-UHFFFAOYSA-N | |
N-methoxymethanamine;hydrochloride | |
81138 | |
Crystalline Powder or Crystals |
N, O-Dimethylhydroxylamine hydrochloride | |
Glass bottle | |
White | |
5g | |
6638-79-5 | |
100.0 | |
C2H7NO·HCl | |
MFCD00012485 | |
11,201; 12,205; 15,144 | |
(10% in water) Clear | |
CNOC.Cl | |
97.55 | |
97.55 | |
98% |
Chemical Identifiers
6638-79-5 | |
97.55 | |
USZLCYNVCCDPLQ-UHFFFAOYSA-N | |
81138 | |
CNOC.Cl |
C2H7NO·HCl | |
MFCD00012485 | |
methanamine, n-methoxy-, hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylamine hcl, n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride, n-methoxymethanamine hydrochloride, n-methoxymethylamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, o,n-dimethylhydroxylamine hydrochloride | |
N-methoxymethanamine;hydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
EINECSNumber : 229-642-1
TSCA : TSCA