Please login to your online account to display your discounted pricing

N,O-Di-BOC-hydroxylamine, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC427880050

 View more versions of this product

Catalog No. AC427880050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 233.26
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material N, O-Di-BOC-hydroxylamine
MDL Number MFCD00034797
CAS 85006-25-3
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Molecular Formula C10H19NO5
Synonym N, O-di-BOC-hydroxylamine
Packaging Glass bottle
Health Hazard 1 Warning

Specifications

Chemical Name or Material N, O-Di-BOC-hydroxylamine
CAS 85006-25-3
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H19NO5
Formula Weight 233.26
MDL Number MFCD00034797
Synonym N, O-di-BOC-hydroxylamine
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.