Please login to your online account to display your discounted pricing

N,O-Di-BOC-hydroxylamine, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC427880010

 View more versions of this product

Catalog No. AC427880010

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material N, O-Di-BOC-hydroxylamine
CAS 85006-25-3
CAS Min % 96.0
Health Hazard 1 Warning
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 97%
Quantity 1g
Molecular Formula C10H19NO5
Formula Weight 233.26
MDL Number MFCD00034797
Synonym N, O-di-BOC-hydroxylamine

Specifications

Chemical Name or Material N, O-Di-BOC-hydroxylamine
CAS 85006-25-3
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H19NO5
Formula Weight 233.26
MDL Number MFCD00034797
Synonym N, O-di-BOC-hydroxylamine
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.