Please login to your online account to display your discounted pricing

N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC168005000

 View more versions of this product

Catalog No. AC168005000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Liquid
Formula Weight 257.39
Quantity 500g
Color Clear colorless to light yellow
Density 0.9600g/mL
CAS Min % 98.0
CAS Max % 100.0
Linear Formula CF3C[=NSi(CH3)3]OSi(CH3)3
Molecular Formula C8H18F3NOSi2
CAS 25561-30-2
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Refractive Index 1.3829 to 1.3849 (20°C, 589nm)
Packaging Glass bottle
Synonym BSTFA
Chemical Name or Material N, O-Bis(trimethylsilyl)trifluoroacetamide
Assay Percent Range 98+%
MDL Number MFCD00008269
Health Hazard 1 Danger

Specifications

Chemical Name or Material N, O-Bis(trimethylsilyl)trifluoroacetamide
CAS 25561-30-2
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C8H18F3NOSi2
Linear Formula CF3C[=NSi(CH3)3]OSi(CH3)3
Formula Weight 257.39
MDL Number MFCD00008269
Synonym BSTFA
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

SDS & Safety Documents