Learn More
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 10% TMCS, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04017414
Description
N,O-Bis(trimethylsilyl)trifluoroacetamide is a powerful silylation reagent useful in GC and GC-MS for the derivatization of a wide range of functional groups under mild conditions. N,O. It is also used in HPLC- electrochemical detection assay of 8-oxo-deoxyguanosine and?8-oxo-guanine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N,O-Bis(trimethylsilyl)trifluoroacetamide | |
25561-30-2 | |
23°C (73°F) | |
MFCD00008269 | |
UN2924 | |
Moisture sensitive | |
Soluble in water (hydrolyses),and chloroform. | |
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C | |
257.403 | |
257.4 |
with 10% TMCS | |
Colorless to Yellow | |
C8H18F3NOSi2 | |
25 g | |
3607620 | |
bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate | |
XCOBLONWWXQEBS-GHXNOFRVSA-N | |
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate | |
9601896 | |
Liquid |
Chemical Identifiers
25561-30-2 | |
257.403 | |
XCOBLONWWXQEBS-GHXNOFRVSA-N | |
9601896 | |
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C |
C8H18F3NOSi2 | |
MFCD00008269 | |
bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate | |
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate |
Safety and Handling
GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : Yes
storageNote1 : 2°C to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only