Learn More
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 389320250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagentSpecifications
N, O-Bis(trimethylsilyl)trifluoroacetamide | |
Colorless to Yellow | |
145.0°C to 147.0°C | |
Glass bottle | |
CF3C[=NSi(CH3)3]OSi(CH3)3 | |
25 g | |
bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate | |
XCOBLONWWXQEBS-GHXNOFRVSA-N | |
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate | |
9601896 | |
≥98.0% (excluding TMCS), 1.5% max. TMCS |
25561-30-2,75-77-4 | |
0.9740g/mL | |
34°C | |
C8H18F3NOSi2 | |
MFCD00008269 | |
0.974 | |
Solubility in water: hydrolysis | |
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C | |
257.4 | |
257.4 | |
Liquid |
Chemical Identifiers
25561-30-2 | |
257.4 | |
XCOBLONWWXQEBS-GHXNOFRVSA-N | |
9601896 | |
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C |
C8H18F3NOSi2 | |
MFCD00008269 | |
bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate | |
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It i
GHS Signal Word: Danger
EINECSNumber : 247-103-9
RUO – Research Use Only