missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS

Manufacturer:  Alfa Aesar™ 4393946

 View more versions of this product

Catalog No. AA4393946



Description

Description

  • Ampouled under argon
Specifications

Specifications

N,O-Bis (trimethylsilyl)trifluoroacetamide
23°C (73°F)
1.3840
145°C to 147°C
-10°C
With 1% TMCS
C8H18F3NOSi2
UN2924
bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
257.403
257.4
0.97g/mL
Ampule
Moisture Sensitive
Colorless to Yellow
5 x 1g
25561-30-2
MFCD00008269
3607620
XCOBLONWWXQEBS-GHXNOFRVSA-N
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
9601896
Liquid

Chemical Identifiers

25561-30-2
257.403
XCOBLONWWXQEBS-GHXNOFRVSA-N
9601896
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
C8H18F3NOSi2
MFCD00008269
bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Recommended Storage : 2°C to 8°C

Documents