Learn More
n-Nonylamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 129181000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Nonylamine | |
97.5 | |
-1.0°C | |
0.7840g/mL | |
63°C | |
97.5% min. (GC) | |
C9H22ClN | |
CH3(CH2)8NH2 | |
100 mL | |
0.784 | |
Solubility in water: slightly soluble | |
[Cl-].CCCCCCCCC[NH3+] | |
179.73 | |
143.27 | |
Liquid |
112-20-9 | |
100.0 | |
Colorless to Yellow | |
202.0°C | |
Authentic | |
Glass bottle | |
1.4320 to 1.4340 | |
MFCD00008249 | |
04,198 | |
nonylamine, n-nonylamine, 1-aminononane, 1-nonanamine, 1-nonylamine, unii-7l7h2ht4ok, 7l7h2ht4ok, nonyl-amine, n-nonyl amine, amine c9 | |
IMGHCWMUWVHYCO-UHFFFAOYSA-N | |
nonan-1-amine | |
16215 | |
98% |
Chemical Identifiers
112-20-9 | |
179.73 | |
IMGHCWMUWVHYCO-UHFFFAOYSA-N | |
16215 | |
[Cl-].CCCCCCCCC[NH3+] |
C9H22ClN | |
MFCD00008249 | |
nonylamine, n-nonylamine, 1-aminononane, 1-nonanamine, 1-nonylamine, unii-7l7h2ht4ok, 7l7h2ht4ok, nonyl-amine, n-nonyl amine, amine c9 | |
nonan-1-amine |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED
GHS Signal Word: Danger
EINECSNumber : 203-945-9
RUO – Research Use Only