missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Nonane, 99%, pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 129110020

 View more versions of this product

Catalog No. AC129110020


 

Specifications Description & Specifications

Specifications

n-Nonane
Clear colorless
111-84-2
100.0
Glass bottle
Pure
≥98.5%
C9H20
128.26
01, 165
0.7180g/mL
2mL
98.5
Authentic
1.4040 to 1.4060 (20°C, 589nm)
Liquid
99%
CH3(CH2)7CH3
MFCD00009574

Chemical Identifiers

111-84-2
100.0
C9H20
128.26
n-Nonane
Pure
98.5
99%
CH3(CH2)7CH3
MFCD00009574
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause long lasting harmful effects to aquatic life.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger