missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N′-Bis(acryloyl)cystamine, 98%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4413203

 View more versions of this product

Catalog No. AA4413203



Specifications

Specifications

N, N′-Bis(acryloyl)cystamine
1g
98%
MFCD00036225
Soluble in acetic acid. Slightly soluble in water.
C=CC(=O)NCCSSCCNC(=O)C=C
260.37
260.37
123°C to 124°C
60984-57-8
C10H16N2O2S2
2,2'-bisacrylamino diethyldisulfide
DJVKJGIZQFBFGS-UHFFFAOYSA-N
N-[2-[2-(prop-2-enoylamino)ethyldisulfanyl]ethyl]prop-2-enamide
100602
Powder, Packaged Under Argon

Chemical Identifiers

60984-57-8
260.37
DJVKJGIZQFBFGS-UHFFFAOYSA-N
100602
C=CC(=O)NCCSSCCNC(=O)C=C
C10H16N2O2S2
MFCD00036225
2,2'-bisacrylamino diethyldisulfide
N-[2-[2-(prop-2-enoylamino)ethyldisulfanyl]ethyl]prop-2-enamide
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 262-546-8

TSCA : No

Recommended Storage : Store at -20°C; Store under Nitrogen

Documents