Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(n-Propyl)maleimide, 94%

Chemical Identifiers

Chemical Name or Material N-(n-Propyl)maleimide
CAS 21746-40-7
Molecular Formula C7H9NO2
Formula Weight 139.15
MDL Number MFCD00014541
UN Number UN2811
Beilstein 1525008
Synonym 1-(n-Propyl)-1H-pyrrole-2,5-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0046006

 
alfa aesar™
L0046006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0046014

 
alfa aesar™
L0046014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 139.15
Synonym 1-(n-Propyl)-1H-pyrrole-2,5-dione
Health Hazard 3 GHS P Statement
P280d-P305+P351+P338-P309-P310
Wear protective clothing and eye protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material N-(n-Propyl)maleimide
Molecular Formula C7H9NO2
MDL Number MFCD00014541
CAS 21746-40-7
Health Hazard 2 GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Melting Point 24° to 26°C
Boiling Point 97°C (18mmHg)
Assay 94%
Quantity 5g
Flash Point 88°C (190°F)
UN Number UN2811
Beilstein 1525008
Health Hazard 1 Danger
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Safety and Handling

Danger

GHS P Statement
P280d-P305+P351+P338-P309-P310
Wear protective clothing and eye protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

2° to 8°C