Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(n-Propyl)ethylenediamine, 97%

Chemical Identifiers

Chemical Name or Material N-(n-Propyl)ethylenediamine
CAS 111-39-7
Molecular Formula C5H14N2
Formula Weight 102.18
MDL Number MFCD00008172
UN Number UN2733
Beilstein 1697232
Synonym 2-(n-Propylamino)ethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0351704

  SDS
alfa aesar™
L0351704
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0351709

  SDS
alfa aesar™
L0351709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 102.18
Chemical Name or Material N-(n-Propyl)ethylenediamine
MDL Number MFCD00008172
Synonym 2-(n-Propylamino)ethylamine
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
TSCA TSCA
Boiling Point 147° to 150°C
Assay 97%
Quantity 2g
Molecular Formula C5H14N2
CAS 111-39-7
UN Number UN2733
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 45°C (113°F)
Beilstein 1697232
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.