missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-n-Octylformamide 98.0+%, TCI America™

Manufacturer:  TCI America O037925G

 View more versions of this product

Catalog No. O037925G



Specifications

Specifications

N-n-Octylformamide
Yellow
6282-06-0
ZBWPKQRQZDZVSF-UHFFFAOYSA-N
N-octylformamide
221394
Clear Liquid at 20°C
291°C
25g
C9H19NO
CCCCCCCCNC=O
157.257
157.26
≥98.0% (GC)

Chemical Identifiers

6282-06-0
157.257
221394
CCCCCCCCNC=O
C9H19NO
ZBWPKQRQZDZVSF-UHFFFAOYSA-N
N-octylformamide
Safety and Handling

Safety and Handling

TSCA : No

Documents