Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(n-Octyl)-n-pentylamine, 98+%

Chemical Identifiers

Chemical Name or Material N-(n-Octyl)-n-pentylamine
CAS 6835-13-8
Molecular Formula C13H29N
Formula Weight 199.38
MDL Number MFCD03093075
UN Number UN2735
Beilstein 7524567
Synonym N-(n-Amyl)-n-octylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1886903

  SDS
alfa aesar™
L1886903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL18869MD

  SDS
alfa aesar™
L18869MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.38
Chemical Name or Material N-(n-Octyl)-n-pentylamine
Molecular Formula C13H29N
Assay ≥98%
CAS 6835-13-8
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
MDL Number MFCD03093075
Synonym N-(n-Amyl)-n-octylamine
Beilstein 7524567
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2735
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.