missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-n-Octyl-D-glucamine, 98%

Manufacturer:  Alfa Aesar™ H60709MD

 View more versions of this product

Catalog No. AAH60709MD



Specifications

Specifications

−15° (c=1 in Methanol)
250mg
C14H31NO5
1-deoxy-1-n-octylamino-d-glucitol, 1-deoxy-1-octylamine-d-glucitol, 1-deoxy-1-octylamino-d-glucitol, 1-octylamino-1-deoxy-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentaol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentol, d-glucitol, 1-deoxy-1-octylamino, n-n-octyl-d-glucamine, n-octyl-d-glucamine, n-octylglucamine
CCCCCCCCNCC(C(C(C(CO)O)O)O)O
293.404
293.4
121°C to 124°C
23323-37-7
MFCD00134352
ZRRNJJURLBXWLL-REWJHTLYSA-N
(2R,3R,4R,5S)-6-(octylamino)hexane-1,2,3,4,5-pentol
90064
98%

Chemical Identifiers

23323-37-7
293.404
ZRRNJJURLBXWLL-REWJHTLYSA-N
90064
CCCCCCCCNCC(C(C(C(CO)O)O)O)O
C14H31NO5
MFCD00134352
1-deoxy-1-n-octylamino-d-glucitol, 1-deoxy-1-octylamine-d-glucitol, 1-deoxy-1-octylamino-d-glucitol, 1-octylamino-1-deoxy-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentaol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentol, d-glucitol, 1-deoxy-1-octylamino, n-n-octyl-d-glucamine, n-octyl-d-glucamine, n-octylglucamine
(2R,3R,4R,5S)-6-(octylamino)hexane-1,2,3,4,5-pentol
Safety and Handling

Safety and Handling

EINECSNumber : 245-582-9

TSCA : No

Recommended Storage : Ambient temperatures

Documents