missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,O-Tris(trimethylsilyl)hydroxylamine, TCI America™

Manufacturer:  TCI America T16755G

 View more versions of this product

Catalog No. T16755G



Specifications

Specifications

N,N,O-Tris(trimethylsilyl)hydroxylamine
Yellow
5g
C9H27NOSi3
1760
C[Si](C)(C)N(O[Si](C)(C)C)[Si](C)(C)C
249.576
249.58
≥97.0% (GC)
128°C
-36°C
21023-20-1
MFCD01321190
WEFDIXZMPUWSQQ-UHFFFAOYSA-N
[dimethyl-[trimethylsilyl(trimethylsilyloxy)amino]silyl]methane
519835
Clear Liquid at 20°C

Chemical Identifiers

21023-20-1
249.576
WEFDIXZMPUWSQQ-UHFFFAOYSA-N
[dimethyl-[trimethylsilyl(trimethylsilyloxy)amino]silyl]methane
C9H27NOSi3
MFCD01321190
519835
C[Si](C)(C)N(O[Si](C)(C)C)[Si](C)(C)C
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents