Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N',N''-Triphenylguanidine, 97%

$55.55 - $738.85

Chemical Identifiers

Chemical Name or Material N,N',N''-Triphenylguanidine
CAS 101-01-9
Molecular Formula C19H17N3
Formula Weight 287.36
MDL Number MFCD00020648
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1879406

 
alfa aesar™
A1879406
5g Each for $55.55

AAA1879414

 
alfa aesar™
A1879414
25g Each for $220.12

AAA1879422

 
alfa aesar™
A1879422
100g Each for $738.85
Description & Specifications

Specifications

Formula Weight 287.36
Chemical Name or Material N,N',N''-Triphenylguanidine
TSCA TSCA
Melting Point 142° to 145°C
Assay 97%
Quantity 5g
Molecular Formula C19H17N3
MDL Number MFCD00020648
CAS 101-01-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number MF6825000

Applications
It is employed as a intermediate for organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.