Please login to your online account to display your discounted pricing

N,N,N'-Trimethylethylenediamine, 98%, Alfa Aesar™

Used as a precursor in the synthesis of a novel gemini surfactant viz. N,N-dimethyl-N-{2-[N'-methyl-N'-(3-sulfopropyl)-alkylammonium]ethyl}-1-alkylammonium bromides. Also used as an amine component for in-situ formation of alpha-amino alkoxides. Involved in the synthesis of N,N'-dimethyl-N'-(2-hydroxy-aethyl)-aethylendiamin.

$283.27 - $283.27

Chemical Identifiers

Chemical Name or Material N,N,N'-Trimethylethylenediamine
CAS 142-25-6
Linear Formula (CH3)2NCH2CH2NHCH3
Formula Weight 102.18
MDL Number MFCD00014874
UN Number UN2733
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3178514

  SDS
alfa aesar
3178514
25g Each for $283.27

AA3178503

  SDS
alfa aesar
3178503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3178506

  SDS
alfa aesar
3178506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Molecular Formula C5H14N2
Formula Weight 102.18
EINECS Number 205-529-2
Storage Note 1 Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
UN Number UN2733
TSCA TSCA
Density 0.804g/mL
MDL Number MFCD00014874
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Flash Point 9° (49°F)
Odor Amine-like
Physical Form Liquid
Quantity 25g
Assay 98%
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
CAS 142-25-6
Refractive Index 1.4190
Boiling Point 116° to 118°C
Chemical Name or Material N,N,N'-Trimethylethylenediamine
Linear Formula (CH3)2NCH2CH2NHCH3
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.