missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N′-Trimethyl-N′-(2-hydroxyethyl)bis(2-aminoethyl) Ether, TCI America™

Manufacturer:  TCI America T159825ML

 View more versions of this product

Catalog No. T159825ML



Specifications

Specifications

N,N,N′-Trimethyl-N′-(2-hydroxyethyl)bis(2-aminoethyl) Ether
25mL
C9H22N2O2
2735
NCUPDIHWMQEDPR-UHFFFAOYSA-N
2-[2-[2-(dimethylamino)ethoxy]ethyl-methylamino]ethanol
3086157
Clear Liquid at 20°C
Yellow
83016-70-0
MFCD00671578
N-(2-Hydroxyethyl)-N,N′C,N′C-trimethylbis(2-aminoethyl) Ether, 2-[[2-[2-(Dimethylamino)ethoxy]ethyl]methylamino]ethanol
CN(C)CCOCCN(C)CCO
190.287
190.29
≥98.0% (GC,T)

Chemical Identifiers

83016-70-0
190.287
NCUPDIHWMQEDPR-UHFFFAOYSA-N
3086157
CN(C)CCOCCN(C)CCO
C9H22N2O2
MFCD00671578
N-(2-Hydroxyethyl)-N,N′C,N′C-trimethylbis(2-aminoethyl) Ether, 2-[[2-[2-(Dimethylamino)ethoxy]ethyl]methylamino]ethanol
2-[2-[2-(dimethylamino)ethoxy]ethyl-methylamino]ethanol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-4166

TSCA : Yes

Documents