Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N,N'-Trimethyl-1,3-propanediamine, 97%, stab. with KOH

Chemical Identifiers

Chemical Name or Material N,N,N'-Trimethyl-1,3-propanediamine
CAS 4543-96-8
Name Note stabilized with KOH
Molecular Formula (CH3)2NCH2CH2CH2NHCH3
Formula Weight 116.21
MDL Number MFCD00014879
UN Number UN2734
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1766014

  SDS
alfa aesar™
A1766014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1766022

  SDS
alfa aesar™
A1766022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material N,N,N'-Trimethyl-1,3-propanediamine
Name Note stabilized with KOH
CAS 4543-96-8
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
TSCA TSCA
Boiling Point 140° to 142°C
Assay 97%
Quantity 25g
Flash Point 34°C (94°F)
Molecular Formula (CH3)2NCH2CH2CH2NHCH3
Formula Weight 116.21
MDL Number MFCD00014879
UN Number UN2734

Applications
N,N,N′-Trimethyl-1,3-propanediamine is employed in the preparation of unlabelled N,N,N′-trimethyl-[2-hydroxy-3-methyl-5-iodobenzyl]-1,3-propanediamine (HIDPM).

Solubility
Soluble in water.

Notes
Air Sensitive. Store away from acids, air and oxidizing agents. Store under inert gas.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.