missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N′,N′-Tetraphenylbenzidine, TCI America™

Manufacturer:  TCI America T18125G

 View more versions of this product

Catalog No. T18125G



Specifications

Specifications

N,N,N′,N′-Tetraphenylbenzidine
233°C
15546-43-7
MFCD00228123
DCZNSJVFOQPSRV-UHFFFAOYSA-N
N,N-diphenyl-4-[4-(N-phenylanilino)phenyl]aniline
84981
Crystal-Powder at 20°C
White-Yellow
5g
C36H28N2
n,n,n',n'-tetraphenylbenzidine, tetraphenylbenzidine, n4,n4,n4',n4'-tetraphenyl-1,1'-biphenyl-4,4'-diamine, n,n,n,n-tetraphenylbenzidine, 1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetraphenyl, n,n'-diphenyl-n,n',di-m-tolyl-benzidine, n,n,n',n'-tetraphenyl-1,1'-biphenyl-4,4'-diamine, 1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetraphenyl, 4,4'-bis diphenylamino biphenyl, 1,1'-biphenyl-4,4'-diamine, n4,n4,n4',n4'-tetraphenyl
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)N(C5=CC=CC=C5)C6=CC=CC=C6
488.634
488.63
≥98.0% (GC)

Chemical Identifiers

15546-43-7
488.634
DCZNSJVFOQPSRV-UHFFFAOYSA-N
84981
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)N(C5=CC=CC=C5)C6=CC=CC=C6
C36H28N2
MFCD00228123
n,n,n',n'-tetraphenylbenzidine, tetraphenylbenzidine, n4,n4,n4',n4'-tetraphenyl-1,1'-biphenyl-4,4'-diamine, n,n,n,n-tetraphenylbenzidine, 1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetraphenyl, n,n'-diphenyl-n,n',di-m-tolyl-benzidine, n,n,n',n'-tetraphenyl-1,1'-biphenyl-4,4'-diamine, 1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetraphenyl, 4,4'-bis diphenylamino biphenyl, 1,1'-biphenyl-4,4'-diamine, n4,n4,n4',n4'-tetraphenyl
N,N-diphenyl-4-[4-(N-phenylanilino)phenyl]aniline
Safety and Handling

Safety and Handling

TSCA : Yes

Documents