Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N,N',N'-Tetramethylsulfonamide, 98%

$76.05 - $76.05

Chemical Identifiers

Chemical Name or Material N,N,N',N'-Tetramethylsulfonamide
CAS 3768-63-6
Molecular Formula C4H12N2O2S
Formula Weight 152.22
MDL Number MFCD00043628
Beilstein 1759783
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0913303

 
alfa aesar™
L0913303
1g Each for $76.05
Description & Specifications

Specifications

Formula Weight 152.22
Chemical Name or Material N,N,N',N'-Tetramethylsulfonamide
Melting Point 70° to 73°C
Assay 98%
MDL Number MFCD00043628
CAS 3768-63-6
Beilstein 1759783
Quantity 1g
Molecular Formula C4H12N2O2S