Please login to your online account to display your discounted pricing

N,N,N',N'-Tetramethylbenzidine 97.5%, ACROS Organics™

$275.54 - $275.54

Chemical Identifiers

Chemical Name or Material N,N,N',N'-Tetramethylbenzidine
Name Note 97.5%
CAS 366-29-0
CAS Min % 96.5
CAS Max % 100.0
Assay ≥96.5%
Assay Percent Range 97.50%
Molecular Formula C16H20N2
Linear Formula ((CH3)2NC6H4)2
Formula Weight 240.35
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC138380050

  SDS
fisher scientific acros
138380050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC138380250

  SDS
thermo fisher scientific
138380250
Glass bottle 25g Each for $275.54
Description & Specifications

Specifications

Assay ≥96.5%
Color Brownish
Formula Weight 240.35
Physical Form Fine Powder
Quantity 5g
Beilstein 13, 221
TSCA TSCA
EINECS Number 206-676-5
RTECS Number DD1752100
Melting Point 192° to 196°C
CAS Max % 100.0
CAS Min % 96.5
Packaging Glass bottle
Infrared Spectrum Authentic
MDL Number MFCD00008310
Chemical Name or Material N,N,N',N'-Tetramethylbenzidine
CAS 366-29-0
Name Note 97.5%
Assay Percent Range 97.50%
Linear Formula ((CH3)2NC6H4)2
Molecular Formula C16H20N2