Learn More
N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiuronium hexafluorophosphate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC440260250
Description
Coupling reagentSpecifications
N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiuronium hexafluorophosphate | |
Authentic | |
111.7°C to 113.6°C | |
212333-72-7 | |
100.0 | |
C10H16N3OS·F6P | |
UCOGEMMJHLHOAW-UHFFFAOYSA-N | |
[dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;hexafluorophosphate | |
10571248 | |
Powder |
30.41 to 34.29% C, 10.64 to 12.00% N | |
White to Yellow | |
25g | |
96.0 | |
97% | |
acmc-209fij, ambotzrl-1156, hott, hott n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, ksc911c8f, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuron, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronhexafluorophosphate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, n,n,n,n-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate | |
CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-].F[P-](F)(F)(F)(F)F | |
371.29 | |
371.29 | |
97% |
Chemical Identifiers
212333-72-7 | |
371.29 | |
acmc-209fij, ambotzrl-1156, hott, hott n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, ksc911c8f, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuron, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronhexafluorophosphate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, n,n,n,n-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate | |
[dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;hexafluorophosphate |
C10H16N3OS·F6P | |
UCOGEMMJHLHOAW-UHFFFAOYSA-N | |
10571248 | |
CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-].F[P-](F)(F)(F)(F)F |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning