Please login to your online account to display your discounted pricing

N,N,N',N'-Tetramethyl-p-phenylenediamine 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  170790250

 View more versions of this product

Catalog No. AC170790250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Flakes
Assay ≥97.5% (total base)
Formula Weight 164.25
Quantity 25g
Color Slightly brown to beige-brown
Melting Point 48° to 52°C
Linear Formula C6H4N(CH3)2!2
CAS 100-22-1
Chemical Name or Material N,N,N',N'-Tetramethyl-p-phenylenediamine
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C10H16N2

Specifications

Chemical Name or Material N,N,N',N'-Tetramethyl-p-phenylenediamine
Assay ≥97.5% (total base)
Assay Percent Range 98%
Molecular Formula C10H16N2
Linear Formula C6H4N(CH3)2!2
Formula Weight 164.25