missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)uronium Tetrafluoroborate, TCI America™

Manufacturer:  TCI America T22245G

 View more versions of this product

Catalog No. T22245G



Specifications

Specifications

N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)uronium Tetrafluoroborate
195°C
105832-38-0
1759
YEBLHMRPZHNTEK-UHFFFAOYSA-N
[dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;tetrafluoroborate
9857522
Crystal-Powder at 20°C
White
5g
C9H16BF4N3O3
tstu, o-n-succinimidyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-2,5-dioxopyrrolidin-1-yl-1,1,3,3-tetramethylisouronium tetrafluoroborate, 2-succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl-uronium tetrafluoroborate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl uronium bf4, pubchem12781
[B-](F)(F)(F)F.CN(C)C(=[N+](C)C)ON1C(=O)CCC1=O
301.049
301.05
≥98.0% (HPLC,N)

Chemical Identifiers

105832-38-0
301.049
tstu, o-n-succinimidyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-2,5-dioxopyrrolidin-1-yl-1,1,3,3-tetramethylisouronium tetrafluoroborate, 2-succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl-uronium tetrafluoroborate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl uronium bf4, pubchem12781
[dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;tetrafluoroborate
C9H16BF4N3O3
YEBLHMRPZHNTEK-UHFFFAOYSA-N
9857522
[B-](F)(F)(F)F.CN(C)C(=[N+](C)C)ON1C(=O)CCC1=O
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents