missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440600250

 View more versions of this product

Catalog No. AC440600250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Coupling reagent
Specifications

Specifications

N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate
Authentic
213°C to 215°C
265651-18-1
100.0
C9H16N3O3·F6P
STWZCCVNXFLDDD-UHFFFAOYSA-N
[dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;hexafluorophosphate
16211151
Powder or Crystalline Powder
29.34 to 30.85% C, 11.41 to 11.99% N
White
25g
97.5
98%
2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, ambotzrl-1039, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, hstu, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate
CN(C)C(=[N+](C)C)ON1C(=O)CCC1=O.F[P-](F)(F)(F)(F)F
359.21
359.21
98%

Chemical Identifiers

265651-18-1
359.21
2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, ambotzrl-1039, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, hstu, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate
[dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;hexafluorophosphate
C9H16N3O3·F6P
STWZCCVNXFLDDD-UHFFFAOYSA-N
16211151
CN(C)C(=[N+](C)C)ON1C(=O)CCC1=O.F[P-](F)(F)(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents