missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate, 98%, ACROS Organics™

Chemical Identifiers

CAS 265651-18-1
Molecular Formula C9H16F6N3O3P
Molecular Weight (g/mol) 359.21
MDL Number MFCD01863753
InChI Key STWZCCVNXFLDDD-UHFFFAOYSA-N
Synonym 2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, ambotzrl-1039, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, hstu, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate
PubChem CID 16211151
IUPAC Name [(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC440600250
SDS View Documents
Thermo Fisher Scientific
AC440600250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC440601000
SDS View Documents
Thermo Fisher Scientific
AC440601000
100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Coupling reagent
Specifications

Chemical Identifiers

265651-18-1
359.21
STWZCCVNXFLDDD-UHFFFAOYSA-N
16211151
F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C
C9H16F6N3O3P
MFCD01863753
2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, ambotzrl-1039, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, hstu, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate
[(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium; hexafluoro-λ⁵-phosphanuide

Specifications

29.34 to 30.85% C, 11.41 to 11.99% N
White
25g
97.5
98%
MFCD01863753
STWZCCVNXFLDDD-UHFFFAOYSA-N
[(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium; hexafluoro-λ⁵-phosphanuide
16211151
Powder or Crystalline Powder
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate
Authentic
213°C to 215°C
265651-18-1
100.0
C9H16F6N3O3P
2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, ambotzrl-1039, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, hstu, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate
F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C
359.21
359.21
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS