Please login to your online account to display your discounted pricing

N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate, 99%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC440630010

 View more versions of this product

Catalog No. AC440630010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder or Solid
Formula Weight 349.1
Quantity 1g
Color White to yellow
CAS Max % 100.0
Chemical Name or Material N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate
Assay Percent Range 99%
Molecular Formula C12H16N5O2·BF4
CAS 125700-69-8
Synonym O-(4-Oxo-1, 2, 3-benzotriazin-3(4H)-yl)-N
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Elemental Analysis 40.67 to 41.91% C, 19.76 to 20.36% N
Infrared Spectrum Authentic

Specifications

Chemical Name or Material N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate
CAS 125700-69-8
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C12H16N5O2·BF4
Formula Weight 349.1
Synonym O-(4-Oxo-1, 2, 3-benzotriazin-3(4H)-yl)-N
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.