Please login to your online account to display your discounted pricing

N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate
CAS 125700-69-8
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C12H16N5O2·BF4
Formula Weight 349.1
Synonym O-(4-Oxo-1, 2, 3-benzotriazin-3(4H)-yl)-N
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC440630050

  SDS
thermo fisher scientific
AC440630050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC440630010

  SDS
thermo fisher scientific
AC440630010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Solid
Formula Weight 349.1
Quantity 5g
Color White to yellow
CAS Max % 100.0
Synonym O-(4-Oxo-1, 2, 3-benzotriazin-3(4H)-yl)-N
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Chemical Name or Material N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate
CAS 125700-69-8
Molecular Formula C12H16N5O2·BF4
Assay Percent Range 99%
Infrared Spectrum Authentic
Elemental Analysis 40.67 to 41.91% C, 19.76 to 20.36% N
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.