Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N,N',N'-Tetramethyl-O-(2-oxo-1,2-dihydro-1-pyridyl)uroniumtetrafluoroborate, 98%

$66.04 - $192.41

Chemical Identifiers

Chemical Name or Material N,N,N',N'-Tetramethyl-O-(2-oxo-1,2-dihydro-1-pyridyl)uroniumtetrafluoroborate
CAS 125700-71-2
Molecular Formula C10H16BF4N3O2
Formula Weight 297.06
MDL Number MFCD00075475
Synonym 2-(2-Pyridon-1-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate; TPTU
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1949403

  SDS
alfa aesar™
L1949403
1g Each for $66.04

AAL1949406

  SDS
alfa aesar™
L1949406
5g Each for $192.41
Description & Specifications

Specifications

Formula Weight 297.06
Molecular Formula C10H16BF4N3O2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Synonym 2-(2-Pyridon-1-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate; TPTU
Recommended Storage 2° to 8°C
Chemical Name or Material N,N,N',N'-Tetramethyl-O-(2-oxo-1,2-dihydro-1-pyridyl)uroniumtetrafluoroborate
MDL Number MFCD00075475
CAS 125700-71-2
Melting Point ≈140°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C