Learn More
N,N,N',N'-Tetramethyl-1,3-propanediamine, 98+%, Thermo Scientific Chemicals
$43.53 - $305.19
Chemical Identifiers
CAS | 110-95-2 |
---|---|
Molecular Formula | C7H18N2 |
Molecular Weight (g/mol) | 130.235 |
MDL Number | MFCD00008337 |
InChI Key | DMQSHEKGGUOYJS-UHFFFAOYSA-N |
Synonym | n,n,n',n'-tetramethyl-1,3-propanediamine, 1,3-bis dimethylamino propane, tetramethyltrimethylenediamine, n1,n1,n3,n3-tetramethylpropane-1,3-diamine, n,n,n',n'-tetramethyl-1,3-diaminopropane, 1,3-propanediamine, n,n,n',n'-tetramethyl, bis dimethylamino methyl methane, n,n,n',n'-tetramethyltrimethylenediamine, unii-73b9i6hn5r |
PubChem CID | 8084 |
IUPAC Name | N,N,N',N'-tetramethylpropane-1,3-diamine |
SMILES | CN(C)CCCN(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1950009
|
Thermo Scientific Chemicals
A1950009 |
10 g |
Each for $43.53
|
|
|||||
AAA1950018
|
Thermo Scientific Chemicals
A1950018 |
50 g |
Each for $94.67
|
|
|||||
AAA1950030
|
Thermo Scientific Chemicals
A1950030 |
250 g |
Each for $305.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-95-2 | |
130.235 | |
DMQSHEKGGUOYJS-UHFFFAOYSA-N | |
8084 | |
CN(C)CCCN(C)C |
C7H18N2 | |
MFCD00008337 | |
n,n,n',n'-tetramethyl-1,3-propanediamine, 1,3-bis dimethylamino propane, tetramethyltrimethylenediamine, n1,n1,n3,n3-tetramethylpropane-1,3-diamine, n,n,n',n'-tetramethyl-1,3-diaminopropane, 1,3-propanediamine, n,n,n',n'-tetramethyl, bis dimethylamino methyl methane, n,n,n',n'-tetramethyltrimethylenediamine, unii-73b9i6hn5r | |
N,N,N',N'-tetramethylpropane-1,3-diamine |
Specifications
110-95-2 | |
145°C to 146°C | |
C7H18N2 | |
MFCD00008337 | |
UN2733 | |
n,n,n',n'-tetramethyl-1,3-propanediamine, 1,3-bis dimethylamino propane, tetramethyltrimethylenediamine, n1,n1,n3,n3-tetramethylpropane-1,3-diamine, n,n,n',n'-tetramethyl-1,3-diaminopropane, 1,3-propanediamine, n,n,n',n'-tetramethyl, bis dimethylamino methyl methane, n,n,n',n'-tetramethyltrimethylenediamine, unii-73b9i6hn5r | |
CN(C)CCCN(C)C | |
130.235 | |
130.24 | |
N,N,N',N'-Tetramethyl-1,3-propanediamine |
0.82 | |
31°C (87°F) | |
1.4234 | |
10 g | |
1734176 | |
DMQSHEKGGUOYJS-UHFFFAOYSA-N | |
N,N,N',N'-tetramethylpropane-1,3-diamine | |
8084 | |
99% |
Safety and Handling
GHS H Statement
H301-H310-H314-H318-H226-H332
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302-H311+H331-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 203-818-8
RTECSNumber : TX8400000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only