Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N,N',N'-Tetramethyl-1,3-butanediamine, 99%

Chemical Identifiers

Chemical Name or Material N,N,N',N'-Tetramethyl-1,3-butanediamine
CAS 97-84-7
Molecular Formula (CH3)2NCH(CH3)CH2CH2N(CH3)2
Formula Weight 144.26
MDL Number MFCD00025678
UN Number UN2733
Beilstein 1698054
Synonym 1,3-Bis(dimethylamino)butane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1752614

 
alfa aesar™
A1752614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1752606

 
alfa aesar™
A1752606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.26
Chemical Name or Material N,N,N',N'-Tetramethyl-1,3-butanediamine
TSCA TSCA
Boiling Point 164° to 166°C
Assay 99%
Quantity 25g
Flash Point 40°C (105°F)
Molecular Formula (CH3)2NCH(CH3)CH2CH2N(CH3)2
MDL Number MFCD00025678
CAS 97-84-7
UN Number UN2733
Beilstein 1698054
Synonym 1,3-Bis(dimethylamino)butane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H310-H314-H318-H226-H332
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H310-H314-H318-H226-H332
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if inhaled.