Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N,N',N'-Tetrakis(trimethylsilyl)-1,6-hexanediamine, 97%

Chemical Identifiers

Chemical Name or Material N,N,N',N'-Tetrakis(trimethylsilyl)-1,6-hexanediamine
CAS 39772-62-8
Formula Weight 404.93
MDL Number MFCD24848915
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3686909

 
alfa aesar™
H3686909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3686918

 
alfa aesar™
H3686918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 404.93
Chemical Name or Material N,N,N',N'-Tetrakis(trimethylsilyl)-1,6-hexanediamine
Physical Form Liquid
Assay 97%
MDL Number MFCD24848915
CAS 39772-62-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev
Sensitivity Air Sensitive
Boiling Point 110°C (0.05mmHg)
Color Colorless to yellow
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.