Please login to your online account to display your discounted pricing

N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%, ACROS Organics™

$42.40 - $42.40

Chemical Identifiers

Chemical Name or Material N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine
CAS 140-07-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H24N2O4
Linear Formula (HOCH2CH2)2NCH2CH2N(CH2CH2OH)2
Formula Weight 236.31
MDL Number MFCD00002854
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC420480010

  SDS
thermo fisher scientific
AC420480010
Glass bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC420481000

  SDS
fisher scientific acros
AC420481000
Glass bottle 100g Each for $42.40

AC420485000

  SDS
thermo fisher scientific
AC420485000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
pH ≥10 (25% soln.)
Formula Weight 236.31
Physical Form Viscous Liquid
Quantity 1kg
Color Clear colorless to amber
Density 1.1300g/mL
CAS Max % 100.0
Refractive Index 1.4990 to 1.5070 (20°C, 589nm)
Linear Formula (HOCH2CH2)2NCH2CH2N(CH2CH2OH)2
MDL Number MFCD00002854
CAS Min % 98.5
Synonym 2,2',2'',2'''-(Ethylenedinitrilo)tetraethanol
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine
CAS 140-07-8
Molecular Formula C10H24N2O4
Assay Percent Range 99%