missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N′,N′-Tetrakis(2-hydroxyethyl)adipamide, TCI America™

Manufacturer:  TCI America T285925G

 View more versions of this product

Catalog No. T285925G



Specifications

Specifications

N,N,N′,N′-Tetrakis(2-hydroxyethyl)adipamide
128°C
6334-25-4
MFCD07787618
OKRNLSUTBJUVKA-UHFFFAOYSA-N
N,N,N',N'-tetrakis(2-hydroxyethyl)hexanediamide
95283
Crystal-Powder at 20°C
White-Yellow
25g
C14H28N2O6
N,N,N′,N′-Tetrakis(2-hydroxyethyl)hexanediamide
C(CCC(=O)N(CCO)CCO)CC(=O)N(CCO)CCO
320.386
320.39
≥98.0% (GC)

Chemical Identifiers

6334-25-4
320.386
OKRNLSUTBJUVKA-UHFFFAOYSA-N
95283
C(CCC(=O)N(CCO)CCO)CC(=O)N(CCO)CCO
C14H28N2O6
MFCD07787618
N,N,N′,N′-Tetrakis(2-hydroxyethyl)hexanediamide
N,N,N',N'-tetrakis(2-hydroxyethyl)hexanediamide
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3798

TSCA : No

Documents