Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N,N',N'-Tetraethylmethylenediamine, 98%

Chemical Identifiers

Chemical Name or Material N,N,N',N'-Tetraethylmethylenediamine
CAS 102-53-4
Formula Weight 158.29
MDL Number MFCD00015115
UN Number UN2733
Synonym Bis(diethylamino)methane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1129406

 
alfa aesar™
1129406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1129414

 
alfa aesar™
1129414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1129422

 
alfa aesar™
1129422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.29
Chemical Name or Material N,N,N',N'-Tetraethylmethylenediamine
Refractive Index 1.4260
Boiling Point 166° to 169°C
MDL Number MFCD00015115
Health Hazard 1 Danger
TSCA TSCA
Density 0.811g/mL
Physical Form Liquid
Assay 98%
Quantity 5g
Flash Point 42° (108°F)
CAS 102-53-4
UN Number UN2733
Synonym Bis(diethylamino)methane
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.