Please login to your online account to display your discounted pricing

N,N,N',N'-Tetraethylethylenediamine 99+%, ACROS Organics™

$53.11 - $53.11

Chemical Identifiers

Chemical Name or Material N,N,N',N'-Tetraethylethylenediamine
Assay ≥99.0% (GC)
Assay Percent Range 99+%
Molecular Formula C10H24N2
Linear Formula (C2H5)2NCH2CH2N(C2H5)2
Formula Weight 172.31
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC155590100

  SDS
thermo fisher scientific
155590100
Glass bottle 10mL Each for $53.11

AC155591000

  SDS
thermo fisher scientific
155591000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC155595000

  SDS
thermo fisher scientific
AC155595000
Glass bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0% (GC)
Formula Weight 172.31
Physical Form Liquid
Quantity 10mL
Color Clear light yellow
Density 0.808g/mL
Chemical Name or Material N,N,N',N'-Tetraethylethylenediamine
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4333 to 1.4353 (20°C, 589nm)
Assay Percent Range 99+%
Molecular Formula C10H24N2
Linear Formula (C2H5)2NCH2CH2N(C2H5)2
CAS 150-77-6