Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N,N',N'-Tetraethylethylenediamine, 99%

$193.11 - $193.11

Chemical Identifiers

Chemical Name or Material N,N,N',N'-Tetraethylethylenediamine
CAS 150-77-6
Molecular Formula C10H24N2
Formula Weight 172.32g/mol
MDL Number MFCD00009055
UN Number UN2734
Beilstein 1738741
Synonym 1,2-Bis(diethylamino)ethane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1836214

 
alfa aesar™
A18362-14
25g Each for $193.11
Description & Specifications

Specifications

Boiling Point 191° to 192°C
Quantity 25g
Flash Point 58°C (136°F)
Odor Amine-like
Chemical Name or Material N,N,N',N'-Tetraethylethylenediamine
CAS 150-77-6
Assay 99%
Molecular Formula C10H24N2
Formula Weight 172.32g/mol
MDL Number MFCD00009055
UN Number UN2734
Beilstein 1738741
Synonym 1,2-Bis(diethylamino)ethane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.