Please login to your online account to display your discounted pricing

N,N,N',N'-Tetraethylethylenediamine, 98%, ACROS Organics™

$133.42 - $133.42

Chemical Identifiers

Chemical Name or Material N,N,N';,N';-Tetraethylethylenediamine
CAS 150-77-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H24N2
Linear Formula (C2H5)2NCH2CH2N(C2H5)2
Formula Weight 172.31
MDL Number MFCD00009055
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC293421000

  SDS
thermo fisher scientific
AC293421000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC293420250

  SDS
thermo fisher scientific
293420250
Glass bottle 25mL Each for $133.42
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 172.31
Quantity 100mL
Color Clear light yellow
Density 0.8080g/mL
Beilstein 04, 251
CAS Max % 100.0
Chemical Name or Material N,N,N';,N';-Tetraethylethylenediamine
Refractive Index 1.4333 to 1.4353 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C10H24N2
MDL Number MFCD00009055
CAS 150-77-6
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (C2H5)2NCH2CH2N(C2H5)2
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.