Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N,N',N'-Tetraacetylethylenediamine, 99%

$68.86 - $198.89

Chemical Identifiers

Chemical Name or Material N,N,N',N'-Tetraacetylethylenediamine
CAS 10543-57-4
Molecular Formula C10H16N2O4
Formula Weight 228.25
MDL Number MFCD00014967
Beilstein 1795711
Merck Index 14,9028
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0435318

  SDS
alfa aesar™
L0435318
50g Each for $68.86

AAL0435330

  SDS
alfa aesar™
L0435330
250g Each for $198.89
Description & Specifications

Specifications

Formula Weight 228.25
Chemical Name or Material N,N,N',N'-Tetraacetylethylenediamine
MDL Number MFCD00014967
CAS 10543-57-4
TSCA TSCA
Melting Point 149° to 153°C
Assay 99%
Quantity 50g
Molecular Formula C10H16N2O4
Beilstein 1795711
Merck Index 14,9028

Applications
N,N,N′,N′-Tetraacetylethylenediamine is used as a peroxide bleach activator in household detergents and for paper pulp. It also serves as an important component of laundry detergents and bleaches, where it is used as an activator for active oxygen bleaching agents such as sodium perborate, sodium percarbonate, sodium perphosphate and sodium persulfate. It reacts with the perhydroxyl anion HO2- in the presence of aqueous alkaline solution to prepare triacetylethylenediamine and diacetylethylenediamine with the release of peracetic acid, which is a fast acting bleaching agent.

Solubility
Soluble in chloroform and methylenechloride. Slightly soluble in water and acetone.

Notes
Incompatible with bases and metals.