missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N″,N″-Pentamethyldiethylenetriamine 99.0+%, TCI America™
Supplier: TCI America P0881500ML
Specifications
| N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine | |
| -20°C | |
| 86°C | |
| MFCD00014876 | |
| 2922 | |
| UKODFQOELJFMII-UHFFFAOYSA-N | |
| N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine | |
| 18196 | |
| 173.30 | |
| Liquid |
| 3030-47-5 | |
| Yellow | |
| C9H23N3 | |
| 500 mL | |
| pentamethyldiethylenetriamine, 1,1,4,7,7-pentamethyldiethylenetriamine, pmdt, n,n,n',n,n-pentamethyldiethylenetriamine, pmdeta, pmdta, pentamethyldiethylenetriaminek, bis 2-dimethylaminoethyl methyl amine, 2,5,8-trimethyl-2,5,8-triazanonane, n,n',n-pentamethyldiethylenetriamine | |
| CN(C)CCN(C)CCN(C)C | |
| 173.304 | |
| CHEBI:39475 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 3030-47-5 | |
| 173.304 | |
| UKODFQOELJFMII-UHFFFAOYSA-N | |
| 18196 | |
| N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine |
| C9H23N3 | |
| MFCD00014876 | |
| pentamethyldiethylenetriamine, 1,1,4,7,7-pentamethyldiethylenetriamine, pmdt, n,n,n',n,n-pentamethyldiethylenetriamine, pmdeta, pmdta, pentamethyldiethylenetriaminek, bis 2-dimethylaminoethyl methyl amine, 2,5,8-trimethyl-2,5,8-triazanonane, n,n',n-pentamethyldiethylenetriamine | |
| CHEBI:39475 | |
| CN(C)CCN(C)CCN(C)C |
Safety and Handling
EINECSNumber : (2)-0147
RTECSNumber : IE2100000
TSCA : Yes