missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N′,N′-Ethylenediaminetetrakis(methylenephosphonic Acid), TCI America™

Manufacturer:  TCI America E0393100G

 View more versions of this product

Catalog No. E0393100G



Specifications

Specifications

N,N,N′,N′-Ethylenediaminetetrakis(methylenephosphonic Acid)
100g
1429-50-1
MFCD00058893
NFDRPXJGHKJRLJ-UHFFFAOYSA-N
[2-[bis(phosphonomethyl)amino]ethyl-(phosphonomethyl)amino]methylphosphonic acid
15025
Crystal-Powder at 20°C
White
<2 (1% H2O soln. 25°C)
C6H20N2O12P4
edtmp, edtpo, ethylenebis nitrilodimethylene tetraphosphonic acid, unii-v4cp8rsx7v, v4cp8rsx7v, ethylenediamine tetrakis methylene phosphonic acid, ethane-1,2-diylbis azanetriyl tetrakis methylene tetraphosphonic acid, ethylenedinitrilo-tetramethylenephosphonic acid, ethylenediamine tetramethylenephosphonic acid, n,n,n',n'-ethylenediaminetetrakis methylenephosphonic acid
C(CN(CP(=O)(O)O)CP(=O)(O)O)N(CP(=O)(O)O)CP(=O)(O)O
436.123
436.12
≥98.0% (T)

Chemical Identifiers

1429-50-1
436.123
NFDRPXJGHKJRLJ-UHFFFAOYSA-N
15025
C(CN(CP(=O)(O)O)CP(=O)(O)O)N(CP(=O)(O)O)CP(=O)(O)O
C6H20N2O12P4
MFCD00058893
edtmp, edtpo, ethylenebis nitrilodimethylene tetraphosphonic acid, unii-v4cp8rsx7v, v4cp8rsx7v, ethylenediamine tetrakis methylene phosphonic acid, ethane-1,2-diylbis azanetriyl tetrakis methylene tetraphosphonic acid, ethylenedinitrilo-tetramethylenephosphonic acid, ethylenediamine tetramethylenephosphonic acid, n,n,n',n'-ethylenediaminetetrakis methylenephosphonic acid
[2-[bis(phosphonomethyl)amino]ethyl-(phosphonomethyl)amino]methylphosphonic acid
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1873

TSCA : Yes

Documents