missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N'-Methylenebismethacrylamide 96.0+%, TCI America™
$84.60 - $859.37
Chemical Identifiers
CAS | 2359-15-1 |
---|---|
Molecular Formula | C9H14N2O2 |
Molecular Weight (g/mol) | 182.223 |
MDL Number | MFCD00060096 |
InChI Key | TURITJIWSQEMDB-UHFFFAOYSA-N |
PubChem CID | 75381 |
IUPAC Name | 2-methyl-N-[(2-methylprop-2-enoylamino)methyl]prop-2-enamide |
SMILES | CC(=C)C(=O)NCNC(=O)C(=C)C |
Chemical Identifiers
2359-15-1 | |
182.223 | |
TURITJIWSQEMDB-UHFFFAOYSA-N | |
2-methyl-N-[(2-methylprop-2-enoylamino)methyl]prop-2-enamide |
C9H14N2O2 | |
MFCD00060096 | |
75381 | |
CC(=C)C(=O)NCNC(=O)C(=C)C |
Specifications
2359-15-1 | |
C9H14N2O2 | |
25 g | |
CC(=C)C(=O)NCNC(=O)C(=C)C | |
182.223 | |
182.22 | |
Crystalline Powder |
White | |
MFCD00060096 | |
TURITJIWSQEMDB-UHFFFAOYSA-N | |
2-methyl-N-[(2-methylprop-2-enoylamino)methyl]prop-2-enamide | |
75381 | |
≥96.0% (T) | |
N,N′-Methylenebismethacrylamide |
Safety and Handling
TSCA : Yes