Please login to your online account to display your discounted pricing

N,N'-Methylenebisacrylamide, Ultrapure Bioreagent, J.T.Baker™


High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Quantity 100g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  403104

 View more versions of this product

Catalog No. 02003503

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 154.2
CAS 110-26-9
Chemical Name or Material N,N'-Methylenebisacrylamide
Name Note Purified for Electrophoresis
Purity Grade Ultrapure Bioreagent
Molecular Formula CH2(NHCOCH:CH2)2
Packaging Wide Mouth Amber Glass Bottle
Quantity 100g
Specific Gravity 1L = 1.24kg

Specifications

Chemical Name or Material N,N'-Methylenebisacrylamide
CAS 110-26-9
Name Note Purified for Electrophoresis
Purity Grade Ultrapure Bioreagent
Molecular Formula CH2(NHCOCH:CH2)2
Formula Weight 154.2